Assay Details
| Gene Symbol: PARP16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083379.1 | NP_001076848.1 | 1 - 2 | 798 | 58 | |
| GenBank mRNA | BC123725.1 | - | 1 - 2 | 798 | 58 | |
More Information
Set Membership: |
|

PARP16
Genomic Map