Assay Details
| Gene Symbol: COX11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083403.2 | NP_001076872.1 | 2 - 3 | 511 | 63 | |
| GenBank mRNA | BC133444.1 | - | 2 - 3 | 512 | 63 | |
More Information
Set Membership: |
|

COX11
Genomic Map