Assay Details
| Gene Symbol: RABEP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083408.2 | NP_001076877.2 | 3 - 4 | 467 | 66 | |
| GenBank mRNA | BC114873.1 | - | 3 - 4 | 467 | 66 | |
More Information
Set Membership: |
|

RABEP2
Genomic Map