Assay Details
| Gene Symbol: CIAO3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083411.2 | NP_001076880.2 | 3 - 4 | 327 | 68 | |
| GenBank mRNA | BC123768.1 | - | 3 - 4 | 314 | 68 | |
| BT030692.1 | - | 3 - 4 | 327 | 68 | ||
More Information
Set Membership: |
|

CIAO3
Genomic Map