Assay Details
| Gene Symbol: MICU2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083416.1 | NP_001076885.1 | 10 - 11 | 1042 | 83 | |
| GenBank mRNA | BC123699.1 | - | 10 - 11 | 1042 | 83 | |
More Information
Set Membership: |
|

MICU2
Genomic Map