Assay Details
| Gene Symbol: TMEM17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083433.1 | NP_001076902.1 | 3 - 4 | 329 | 126 | |
| GenBank mRNA | BC123468.1 | - | 3 - 4 | 329 | 126 | |
More Information
Set Membership: |
|

TMEM17
Genomic Map