Assay Details
| Gene Symbol: AKT1S1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083434.2 | NP_001076903.1 | 3 - 4 | 657 | 98 | |
| GenBank mRNA | BC133618.1 | - | 3 - 4 | 658 | 98 | |
More Information
Set Membership: |
|

AKT1S1
Genomic Map