Assay Details
| Gene Symbol: ZC2HC1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083437.1 | NP_001076906.1 | 6 - 7 | 640 | 63 | |
| GenBank mRNA | BC114757.1 | - | 6 - 7 | 640 | 63 | |
More Information
Set Membership: |
|

ZC2HC1A
Genomic Map