Assay Details
| Gene Symbol: ABI3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083454.1 | NP_001076923.1 | 7 - 8 | 927 | 60 | |
| GenBank mRNA | BC114719.1 | - | 7 - 8 | 927 | 60 | |
More Information
Set Membership: |
|

ABI3
Genomic Map