Assay Details
| Gene Symbol: EEFSEC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083464.2 | NP_001076933.1 | 4 - 5 | 795 | 67 | |
| GenBank mRNA | BC133603.1 | - | 4 - 5 | 796 | 67 | |
More Information
Set Membership: |
|

EEFSEC
Genomic Map