Assay Details
| Gene Symbol: IL17RB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083467.1 | NP_001076936.1 | 3 - 4 | 871 | 80 | |
| GenBank mRNA | BT029799.1 | - | 3 - 4 | 871 | 80 | |
More Information
Set Membership: |
|

IL17RB
Genomic Map