Assay Details
| Gene Symbol: ACVRL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083479.1 | NP_001076948.1 | 4 - 5 | 632 | 93 | |
| GenBank mRNA | BC123398.1 | - | 4 - 5 | 632 | 93 | |
More Information
Set Membership: |
|

ACVRL1
Genomic Map