Assay Details
| Gene Symbol: CPZ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083486.2 | NP_001076955.1 | 10 - 11 | 1635 | 55 | |
| GenBank mRNA | BC133650.1 | - | 10 - 11 | 1636 | 55 | |
More Information
Set Membership: |
|

CPZ
Genomic Map