Assay Details
| Gene Symbol: DLK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083494.1 | NP_001076963.1 | 1 - 2 | 142 | 120 | |
| GenBank mRNA | BC123884.1 | - | 1 - 2 | 142 | 120 | |
More Information
Set Membership: |
|

DLK2
Genomic Map