Assay Details
| Gene Symbol: GLO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083496.1 | NP_001076965.1 | 1 - 2 | 127 | 67 | |
| GenBank mRNA | BC123482.1 | - | 1 - 2 | 127 | 67 | |
More Information
Set Membership: |
|

GLO1
Genomic Map