Assay Details
| Gene Symbol: ZNF385B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083502.1 | NP_001076971.1 | 6 - 7 | 1129 | 116 | |
| GenBank mRNA | BC123389.1 | - | 6 - 7 | 1129 | 116 | |
More Information
Set Membership: |
|

ZNF385B
Genomic Map