Assay Details
| Gene Symbol: UBE2M | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083503.1 | NP_001076972.1 | 5 - 6 | 921 | 87 | |
| GenBank mRNA | BC133501.1 | - | 5 - 6 | 921 | 87 | |
More Information
Set Membership: |
|

UBE2M
Genomic Map