Assay Details
| Gene Symbol: PDCD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083506.1 | NP_001076975.1 | 1 - 2 | 89 | 132 | |
| GenBank mRNA | AB510901.1 | - | 1 - 2 | 111 | 132 | |
| BC123854.1 | - | 1 - 2 | 89 | 132 | ||
| LC271172.1 | - | 1 - 2 | 67 | 132 | ||
More Information
Set Membership: |
|

PDCD1
Genomic Map