Assay Details
| Gene Symbol: BAALC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083508.1 | NP_001076977.1 | 2 - 3 | 331 | 72 | |
| GenBank mRNA | BC123708.1 | - | 2 - 3 | 331 | 72 | |
More Information
Set Membership: |
|

BAALC
Genomic Map