Assay Details
| Gene Symbol: KRT6A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083510.1 | NP_001076979.1 | 4 - 5 | 945 | 87 | |
| GenBank mRNA | BC123886.1 | - | 4 - 5 | 945 | 87 | |
More Information
Set Membership: |
|

KRT6A
Genomic Map