Assay Details
| Gene Symbol: CERS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083514.2 | NP_001076983.1 | 1 - 2 | 273 | 79 | |
| GenBank mRNA | BC114845.1 | - | 1 - 2 | 274 | 79 | |
More Information
Set Membership: |
|

CERS1
Genomic Map