Assay Details
| Gene Symbol: NDUFAF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083516.1 | NP_001076985.1 | 1 - 2 | 163 | 126 | |
| GenBank mRNA | BC114895.1 | - | 1 - 2 | 163 | 126 | |
More Information
Set Membership: |
|

NDUFAF4
Genomic Map