Assay Details
| Gene Symbol: NDUFAF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083516.1 | NP_001076985.1 | 2 - 3 | 273 | 73 | |
| GenBank mRNA | BC114895.1 | - | 2 - 3 | 273 | 73 | |
More Information
Set Membership: |
|

NDUFAF4
Genomic Map