Assay Details
| Gene Symbol: ZNF34 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083520.1 | NP_001076989.1 | 3 - 4 | 975 | 85 | |
| GenBank mRNA | BC114747.1 | - | 5 - 6 | 813 | 85 | |
| BC133534.1 | - | 3 - 4 | 975 | 85 | ||
More Information
Set Membership: |
|

ZNF34
Genomic Map