Assay Details
| Gene Symbol: UGT8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083635.1 | NP_001077104.1 | 5 - 6 | 1507 | 80 | |
| GenBank mRNA | BC134489.1 | - | 5 - 6 | 1507 | 80 | |
More Information
Set Membership: |
|

UGT8
Genomic Map