Assay Details
| Gene Symbol: PPARD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083636.1 | NP_001077105.1 | 7 - 8 | 1333 | 63 | |
| GenBank mRNA | AF229357.1 | - | 5 - 6 | 1116 | 63 | |
| BC134636.1 | - | 7 - 8 | 1333 | 63 | ||
More Information
Set Membership: |
|

PPARD
Genomic Map