Assay Details
| Gene Symbol: NEU1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083642.1 | NP_001077111.1 | 2 - 3 | 459 | 78 | |
| GenBank mRNA | BC134571.1 | - | 2 - 3 | 459 | 78 | |
More Information
Set Membership: |
|

NEU1
Genomic Map