Assay Details
| Gene Symbol: CDCA8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083652.1 | NP_001077121.1 | 3 - 4 | 323 | 58 | |
| GenBank mRNA | BC134661.1 | - | 3 - 4 | 323 | 58 | |
More Information
Set Membership: |
|

CDCA8
Genomic Map