Assay Details
| Gene Symbol: TIMM23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083658.1 | NP_001077127.1 | 1 - 2 | 206 | 132 | |
| GenBank mRNA | BC134628.1 | - | 1 - 2 | 206 | 132 | |
More Information
Set Membership: |
|

TIMM23
Genomic Map