Assay Details
| Gene Symbol: STMN3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083666.1 | NP_001077135.1 | 1 - 2 | 79 | 64 | |
| GenBank mRNA | BC134627.1 | - | 1 - 2 | 79 | 64 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC134627.1, NM_001083666.1 transcripts.
Set Membership: |
|

STMN3
Genomic Map