Assay Details
| Gene Symbol: SMG5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083679.1 | NP_001077148.1 | 13 - 14 | 2156 | 53 | |
| GenBank mRNA | BC134680.1 | - | 13 - 14 | 2156 | 53 | |
More Information
Set Membership: |
|

SMG5
Genomic Map