Assay Details
| Gene Symbol: PCSK1N | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083680.2 | NP_001077149.1 | 2 - 3 | 651 | 129 | |
| GenBank mRNA | BC134721.1 | - | 2 - 3 | 652 | 129 | |
More Information
Set Membership: |
|

PCSK1N
Genomic Map