Assay Details
| Gene Symbol: CD2BP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083682.2 | NP_001077151.1 | 2 - 3 | 135 | 59 | |
| GenBank mRNA | BC134720.1 | - | 2 - 3 | 136 | 59 | |
More Information
Set Membership: |
|

CD2BP2
Genomic Map