Assay Details
| Gene Symbol: CA8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083690.2 | NP_001077159.1 | 2 - 3 | 591 | 86 | |
| GenBank mRNA | BC134783.1 | - | 2 - 3 | 592 | 86 | |
More Information
Set Membership: |
|

CA8
Genomic Map