Assay Details
| Gene Symbol: TRARG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083707.1 | NP_001077176.1 | 2 - 3 | 795 | 166 | |
| GenBank mRNA | BC134530.1 | - | 2 - 3 | 795 | 166 | |
More Information
Set Membership: |
|

TRARG1
Genomic Map