Assay Details
| Gene Symbol: LMOD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083715.1 | NP_001077184.1 | 1 - 2 | 323 | 88 | |
| GenBank mRNA | BC134619.1 | - | 1 - 2 | 323 | 88 | |
More Information
Set Membership: |
|

LMOD1
Genomic Map