Assay Details
| Gene Symbol: KLF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083727.1 | NP_001077196.1 | 2 - 3 | 1430 | 87 | |
| GenBank mRNA | BC134493.1 | - | 2 - 3 | 1430 | 87 | |
More Information
Set Membership: |
|

KLF5
Genomic Map