Assay Details
| Gene Symbol: GJA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083738.1 | NP_001077207.1 | 1 - 2 | 54 | 95 | |
| GenBank mRNA | BC134629.1 | - | 1 - 2 | 54 | 95 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC134629.1, NM_001083738.1 transcripts.
Set Membership: |
|

GJA4
Genomic Map