Assay Details
| Gene Symbol: TM9SF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083743.1 | NP_001077212.1 | 5 - 6 | 1564 | 74 | |
| GenBank mRNA | BC134551.1 | - | 5 - 6 | 1564 | 74 | |
More Information
Set Membership: |
|

TM9SF1
Genomic Map