Assay Details
| Gene Symbol: SYT5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083744.1 | NP_001077213.1 | 3 - 4 | 335 | 53 | |
| GenBank mRNA | BC134606.1 | - | 3 - 4 | 335 | 53 | |
More Information
Set Membership: |
|

SYT5
Genomic Map