Assay Details
| Gene Symbol: GTF2IRD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083745.1 | NP_001077214.1 | 1 - 2 | 174 | 90 | |
| GenBank mRNA | BC134477.1 | - | 1 - 2 | 174 | 90 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC134477.1, NM_001083745.1 transcripts.
Set Membership: |
|

GTF2IRD2
Genomic Map