Assay Details
| Gene Symbol: GTF2IRD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083745.1 | NP_001077214.1 | 6 - 7 | 753 | 139 | |
| GenBank mRNA | BC134477.1 | - | 6 - 7 | 753 | 139 | |
More Information
Set Membership: |
|

GTF2IRD2
Genomic Map