Assay Details
| Gene Symbol: ZFAND2A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083762.1 | NP_001077231.1 | 6 - 7 | 676 | 92 | |
| GenBank mRNA | BC134789.1 | - | 6 - 7 | 676 | 92 | |
More Information
Set Membership: |
|

ZFAND2A
Genomic Map