Assay Details
| Gene Symbol: IRF8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083769.2 | NP_001077238.1 | 8 - 9 | 1169 | 56 | |
| GenBank mRNA | BC134473.1 | - | 8 - 9 | 1170 | 56 | |
More Information
Set Membership: |
|

IRF8
Genomic Map