Assay Details
| Gene Symbol: CXCL17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083799.2 | NP_001077268.1 | 2 - 3 | 383 | 79 | |
| GenBank mRNA | BC134719.1 | - | 2 - 3 | 385 | 79 | |
More Information
Set Membership: |
|

CXCL17
Genomic Map