Assay Details
| Gene Symbol: GARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097566.1 | NP_001091035.1 | 12 - 13 | 1630 | 147 | |
| GenBank mRNA | AY568575.1 | - | 8 - 9 | 1006 | 147 | |
| BC140482.1 | - | 12 - 13 | 1630 | 147 | ||
More Information
Set Membership: |
|

GARS1
Genomic Map