Assay Details
| Gene Symbol: GARS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097566.1 | NP_001091035.1 | 13 - 14 | 1711 | 101 | |
| GenBank mRNA | AY568575.1 | - | 9 - 10 | 1087 | 101 | |
| BC140482.1 | - | 13 - 14 | 1711 | 101 | ||
More Information
Set Membership: |
|

GARS1
Genomic Map