Assay Details
| Gene Symbol: BLVRA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097571.1 | NP_001091040.1 | 7 - 8 | 708 | 61 | |
| GenBank mRNA | BC140586.1 | - | 7 - 8 | 708 | 61 | |
More Information
Set Membership: |
|

BLVRA
Genomic Map