Assay Details
| Gene Symbol: ALDOC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097984.2 | NP_001091453.1 | 5 - 6 | 621 | 63 | |
| GenBank mRNA | BC103037.1 | - | 5 - 6 | 632 | 63 | |
| BT030500.1 | - | 5 - 6 | 621 | 63 | ||
More Information
Set Membership: |
|

ALDOC
Genomic Map