Assay Details
| Gene Symbol: ANGPT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098855.1 | NP_001092325.1 | 1 - 2 | 635 | 94 | |
| GenBank mRNA | BC142039.1 | - | 1 - 2 | 348 | 94 | |
| BC142471.1 | - | 1 - 2 | 635 | 94 | ||
More Information
Set Membership: |
|

ANGPT2
Genomic Map